Learn More
Medchemexpress LLC Cyclopropylboronic acid | 411235-57-9 | 99.2% | C₃H₇BO₂ | 50 G
SDP
Catalog No. 5000434213
Encompass_Preferred
Cyclopropylboronic acid is an intermediate primarily used in the synthesis of EGFR kinase inhibitors. It plays a role in creating compounds, such as Compound 25, that demonstrate antiproliferative effects on specific EGFR mutant cells, making it valuable for research in lung cancer.
  • Intermediate for EGFR kinase inhibitor synthesis
  • Exhibits antiproliferative effects on EGFR mutant cells
  • Useful in lung cancer research
  • Solid appearance
  • Off-white to yellow color
  • Purity of 99.2%
Catalog No. 50-004-34213 Supplier Medchemexpress LLC Supplier No. HY5921950G
Cyclopropylboronic acid is an intermediate primarily used in the synthesis of EGFR kinase inhibitors. It plays a role in creating compounds, such as Compound 25, that demonstrate antiproliferative effects on specific EGFR mutant cells, making it valuable for research in lung cancer.
  • Intermediate for EGFR kinase inhibitor synthesis
  • Exhibits antiproliferative effects on EGFR mutant cells
  • Useful in lung cancer research
  • Solid appearance
  • Off-white to yellow color
  • Purity of 99.2%