Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclopropylboronic acid | 411235-57-9 | 99.2% | C3H7BO2 | 1 KG
SDP

Catalog No. 5000434147
Encompass_Preferred

Cyclopropylboronic acid is an intermediate used to synthesize EGFR kinase inhibitor (Compound 25). Compound 25 has antiproliferative effects on EGFR mutant (EGFR Δ19del/T790M/C797S) cells and can be used in lung cancer research.

  • Intermediate used to synthesize EGFR kinase inhibitor (Compound 25)
  • Exhibits antiproliferative effects on EGFR mutant (EGFR Δ19del/T790M/C797S) cells
  • Applicable in lung cancer research
  • Inhibits Ba/F3 triple-mutant suspension cells with an IC50 of 642 nM
  • Appearance as an off-white to yellow solid
  • Soluble in DMSO at concentrations of 100 mg/mL or greater
  • Powder stable for 3 years at -20°C
  • In solvent, stable for 6 months at -80°C or 1 month at -20°C

Catalog No. 50-004-34147 Supplier Medchemexpress LLC Supplier No. HY592191KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclopropylboronic acid is an intermediate used to synthesize EGFR kinase inhibitor (Compound 25). Compound 25 has antiproliferative effects on EGFR mutant (EGFR Δ19del/T790M/C797S) cells and can be used in lung cancer research.

  • Intermediate used to synthesize EGFR kinase inhibitor (Compound 25)
  • Exhibits antiproliferative effects on EGFR mutant (EGFR Δ19del/T790M/C797S) cells
  • Applicable in lung cancer research
  • Inhibits Ba/F3 triple-mutant suspension cells with an IC50 of 642 nM
  • Appearance as an off-white to yellow solid
  • Soluble in DMSO at concentrations of 100 mg/mL or greater
  • Powder stable for 3 years at -20°C
  • In solvent, stable for 6 months at -80°C or 1 month at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.