Learn More
Medchemexpress LLC Cyclopropylboronic acid | 411235-57-9 | 99.2% | C3H7BO2 | 5 G
SDP
Catalog No. 5000434148
Encompass_Preferred
Cyclopropylboronic acid is an intermediate used in organic synthesis. It can be utilized to synthesize EGFR kinase inhibitor (Compound 25), which has shown antiproliferative effects on EGFR mutant cells, making it suitable for lung cancer research.
  • Intermediate for organic synthesis
  • Used to synthesize EGFR kinase inhibitor (Compound 25)
  • Compound 25 shows antiproliferative effects on EGFR mutant (EGFR Δ19del/T790M/C797S) cells
  • Applicable in lung cancer research
Catalog No. 50-004-34148 Supplier Medchemexpress LLC Supplier No. HY592195G
Cyclopropylboronic acid is an intermediate used in organic synthesis. It can be utilized to synthesize EGFR kinase inhibitor (Compound 25), which has shown antiproliferative effects on EGFR mutant cells, making it suitable for lung cancer research.
  • Intermediate for organic synthesis
  • Used to synthesize EGFR kinase inhibitor (Compound 25)
  • Compound 25 shows antiproliferative effects on EGFR mutant (EGFR Δ19del/T790M/C797S) cells
  • Applicable in lung cancer research