Learn More
Medchemexpress LLC Cyclopropylboronic acid | 411235-57-9 | 99.22% | C3H7BO2 | 10 G
SDP
Catalog No. 5000434272
Encompass_Preferred
Cyclopropylboronic acid is an intermediate used in the synthesis of EGFR kinase inhibitor (Compound 25). This compound exhibits antiproliferative effects on EGFR mutant (EGFR Δ19del/T790M/C797S) cells. It is also applicable in lung cancer research.
  • Used to synthesize EGFR kinase inhibitor
  • Supports lung cancer research
  • Exhibits antiproliferative effects on EGFR mutant cells
  • For research use only
Catalog No. 50-004-34272 Supplier Medchemexpress LLC Supplier No. HY5921910G
Cyclopropylboronic acid is an intermediate used in the synthesis of EGFR kinase inhibitor (Compound 25). This compound exhibits antiproliferative effects on EGFR mutant (EGFR Δ19del/T790M/C797S) cells. It is also applicable in lung cancer research.
  • Used to synthesize EGFR kinase inhibitor
  • Supports lung cancer research
  • Exhibits antiproliferative effects on EGFR mutant cells
  • For research use only