Learn More
Medchemexpress LLC Cyclopropylboronic acid | 411235-57-9 | MFCD04038750 | 99.2% | C3H7BO2 | 100 G
SDP
Catalog No. 5000433813
Encompass_Preferred
Cyclopropylboronic acid is an intermediate used in the synthesis of EGFR kinase inhibitors, such as Compound 25. This compound exhibits antiproliferative effects on EGFR mutant (EGFR Δ19del/T790M/C797S) cells, making it relevant for lung cancer research.
  • Used to synthesize EGFR kinase inhibitor
  • Exhibits antiproliferative effects on EGFR mutant cells
  • Applicable in lung cancer research
  • Biochemical assay reagent
  • Drug intermediate
Catalog No. 50-004-33813 Supplier Medchemexpress LLC Supplier No. HY59219100G
Cyclopropylboronic acid is an intermediate used in the synthesis of EGFR kinase inhibitors, such as Compound 25. This compound exhibits antiproliferative effects on EGFR mutant (EGFR Δ19del/T790M/C797S) cells, making it relevant for lung cancer research.
  • Used to synthesize EGFR kinase inhibitor
  • Exhibits antiproliferative effects on EGFR mutant cells
  • Applicable in lung cancer research
  • Biochemical assay reagent
  • Drug intermediate