Learn More
Medchemexpress LLC Cyclopropylboronic acid | 411235-57-9 | MFCD04038750 | 99.2% | C3H7BO2 | 500 G
SDP
Catalog No. 5000434002
Encompass_Preferred
Cyclopropylboronic acid is an intermediate that can be utilized in the synthesis of EGFR kinase inhibitor (Compound 25). This compound demonstrates antiproliferative effects on EGFR mutant (EGFR Δ19del/T790M/C797S) cells, suggesting its potential use in lung cancer research.
  • Used to synthesize EGFR kinase inhibitor.
  • Demonstrates antiproliferative effects on EGFR mutant cells.
  • Potential for use in lung cancer research.
  • Inhibits proliferation of Ba/F3 triple-mutant suspension cells.
Catalog No. 50-004-34002 Supplier Medchemexpress LLC Supplier No. HY59219500G
Add to Cart
Add to Cart
Cyclopropylboronic acid is an intermediate that can be utilized in the synthesis of EGFR kinase inhibitor (Compound 25). This compound demonstrates antiproliferative effects on EGFR mutant (EGFR Δ19del/T790M/C797S) cells, suggesting its potential use in lung cancer research.
  • Used to synthesize EGFR kinase inhibitor.
  • Demonstrates antiproliferative effects on EGFR mutant cells.
  • Potential for use in lung cancer research.
  • Inhibits proliferation of Ba/F3 triple-mutant suspension cells.