Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cyclopropylboronic Acid Pinacol Ester

Catalog No. 6598515G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-985-15G 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 65-985-15G Supplier Sigma Aldrich Supplier No. 6598515G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cyclopropylboronic acid pinacol ester can be used:As a cyclopropylating reagent in the study of thiophenol S-cyclopropylation, catalyzed by copper(II) acetate.In one of the key synthetic steps for the preparation of 5-lipoxygenase activating protein (FLAP) inhibitor.

Specifications

CAS 639820-61-4
Melting Point 263°C to 280°C
Molecular Formula C22H31ClN2
Refractive Index n20/D 1.433
Linear Formula C22H31ClN2
MDL Number MFCD08276859
Quantity 500 mg
Synonym 2-Cyclopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol) 358.95 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.