Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopropyldiphenylcarbinol 98.0+%, TCI America™
SDP

Catalog No. C09835G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C09835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C09835G Supplier TCI America Supplier No. C09835G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5785-66-0
Molecular Formula C16H16O
Molecular Weight (g/mol) 224.30
MDL Number MFCD00001298
InChI Key MFKPHBJFWOOEDT-UHFFFAOYSA-N
Synonym cyclopropyldiphenylcarbinol, cyclopropyl diphenyl carbinol, methanol, cyclopropyldiphenyl, alpha-cyclopropylbenzhydrylic alcohol, cyclopropyl diphenyl methanol, diphenyl cyclopropyl carbinol, alpha-cyclopropyl-alpha-phenylbenzyl alcohol, benzyl alcohol, alpha-cyclopropyl-alpha-phenyl, benzenemethanol, alpha-cyclopropyl-alpha-phenyl-9ci, benzenemethanol, alpha-cyclopropyl-alpha-phenyl
PubChem CID 79850
IUPAC Name cyclopropyldiphenylmethanol
SMILES OC(C1CC1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 85°C
Boiling Point 150°C
Quantity 5 g
Formula Weight 224.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Cyclopropyldiphenylcarbinol
RTECSNumber PC1950000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.