Learn More
Medchemexpress LLC Cyclopropylmethyl bromide | 7051-34-5 | 99.9% | 135.00 g/mol | C4H7Br | 100g
SDP
Catalog No. 5000267411
Encompass_Preferred
Cyclopropylmethyl bromide is an intermediate Cyclopropylmethyl bromide can be used in the preparation of Firocoxib (HY-14670) Firocoxib (ML 1785713) is a potent selective orally active COX-2 inhibitor[1]
Catalog No. 50-002-67411 Supplier Medchemexpress LLC Supplier No. HYY1100100G
Cyclopropylmethyl bromide is an intermediate Cyclopropylmethyl bromide can be used in the preparation of Firocoxib (HY-14670) Firocoxib (ML 1785713) is a potent selective orally active COX-2 inhibitor[1]