Learn More
Medchemexpress LLC Cyclopropylmethyl bromide | 7051-34-5 | MFCD00001306 | 99.2% | 135.00 g/mol | C4H7Br | 50g
SDP
Catalog No. 5000267673
Encompass_Preferred
Cyclopropylmethyl bromide is an intermediate Cyclopropylmethyl bromide can be used in the preparation of Firocoxib (HY-14670) Firocoxib (ML 1785713) is a potent selective orally active COX-2 inhibitor[1]
Catalog No. 50-002-67673 Supplier Medchemexpress LLC Supplier No. HYY110050G
Cyclopropylmethyl bromide is an intermediate Cyclopropylmethyl bromide can be used in the preparation of Firocoxib (HY-14670) Firocoxib (ML 1785713) is a potent selective orally active COX-2 inhibitor[1]