Learn More
Medchemexpress LLC Cyclopropylmethyl bromide-d4 | 1219805-93-2 | 97.2% | 139.03 | C4H3D4Br | 1mg
SDP
Supplier:  Medchemexpress LLC HYY1100S1MG
Encompass_Preferred
Cyclopropylmethyl bromide-d4 is the deuterium labeled Cyclopropylmethyl bromide (HY-Y1100) Cyclopropylmethyl bromide is an intermediate Cyclopropylmethyl bromide can be used in the preparation of Firocoxib (HY-14670) Firocoxib (ML 1785713) is a potent selective orally active COX-2 inhibitor[1][2]
Catalog No. 50-002-67407