Learn More
Carbosynth LLC. Cyclorasin 9A5, 1mg, MW: (M.W.1586.86), Molecular formula: C75H108N25O13F
SDP
Supplier:  Carbosynth LLC. IRA3854PI1MG
Encompass_Preferred
Synonyms: cyclo(Trp-Thr-D-Ala-Arg-Arg-Arg-D-2-Nal-Arg-4-F-Phe-D-Nle-Gln), cyclo(WTaRRR-D-2-Nal-R-4-F-Phe-D-Nle-Q), Application: Inhibitor of K-Ras and Lung Cancer Growth. Activates Apoptosis., Storage Temperature: -20°C, References: P. Upadhyay, et al., Angew. Chem. Int. Ed., 54, 1 (2015).
Catalog No. 50-168-5901