Learn More
Medchemexpress LLC Cyclorasin 9A5 (TFA) | 1782098-79-6 | 99.57% | 1586.82 | 5 MG
SDP
Catalog No. 5000423416
Encompass_Preferred
Cyclorasin 9A5 is an 11-residue cell-permeable cyclic peptide that orthosterically inhibits the Ras-Raf protein interaction with an IC50 of 120 nM. It is for research use only and has not been fully validated for medical applications.
  • 11-residue cell-permeable cyclic peptide
  • Orthosterically inhibits Ras-Raf protein interaction
  • IC50 of 120 nM
  • For research use only
Catalog No. 50-004-23416 Supplier Medchemexpress LLC Supplier No. HYP10051A5MG
Cyclorasin 9A5 is an 11-residue cell-permeable cyclic peptide that orthosterically inhibits the Ras-Raf protein interaction with an IC50 of 120 nM. It is for research use only and has not been fully validated for medical applications.
  • 11-residue cell-permeable cyclic peptide
  • Orthosterically inhibits Ras-Raf protein interaction
  • IC50 of 120 nM
  • For research use only