Learn More
Apexbio Technology LLC cyclo(RLsKDK) TFA(Synonyms: cyclo(RLsKDK), Cyclic peptide RLsKDK, ADAM8 inhibitor peptide, Cyclo-Arg-Leu-Ser-Lys-Asp-Lys), 1mg.
SDP
Catalog No. 5000060157
Encompass_Preferred
Cyclo(RLsKDK) TFA is a cyclic pentapeptide inhibitor targeting the metalloproteinase ADAM8 a transmembrane protein frequently dysregulated in pathological conditions such as inflammation cancer and neurodegenerative diseases By specifically inhibiting ADAM8 enzymatic activity cyclo(RLsKDK) TFA may modulate disease-associated cellular processes This molecule holds promise as a research tool for studying ADAM8 function and as a potential therapeutic candidate in inflammatory disorders and oncology
Catalog No. 50-000-60157 Supplier Apexbio Technology LLC Supplier No. A99301
Cyclo(RLsKDK) TFA is a cyclic pentapeptide inhibitor targeting the metalloproteinase ADAM8 a transmembrane protein frequently dysregulated in pathological conditions such as inflammation cancer and neurodegenerative diseases By specifically inhibiting ADAM8 enzymatic activity cyclo(RLsKDK) TFA may modulate disease-associated cellular processes This molecule holds promise as a research tool for studying ADAM8 function and as a potential therapeutic candidate in inflammatory disorders and oncology