Learn More
Medchemexpress LLC Cyclosomatostatin | 84211-54-1 | 99.76% | 779.97 | 5 MG
SDP
Catalog No. 5000424766
Encompass_Preferred
Cyclosomatostatin is a potent somatostatin (SST) receptor antagonist that inhibits somatostatin receptor type 1 (SSTR1) signaling. It has been shown to decrease cell proliferation, ALDH+ cell population size, and sphere-formation in colorectal cancer (CRC) cells.
  • Potent somatostatin receptor antagonist
  • Inhibits somatostatin receptor type 1 (SSTR1) signaling
  • Decreases cell proliferation
  • Reduces ALDH+ cell population size
  • Reduces sphere-formation in colorectal cancer (CRC) cells
Catalog No. 50-004-24766 Supplier Medchemexpress LLC Supplier No. HYP12015MG
Cyclosomatostatin is a potent somatostatin (SST) receptor antagonist that inhibits somatostatin receptor type 1 (SSTR1) signaling. It has been shown to decrease cell proliferation, ALDH+ cell population size, and sphere-formation in colorectal cancer (CRC) cells.
  • Potent somatostatin receptor antagonist
  • Inhibits somatostatin receptor type 1 (SSTR1) signaling
  • Decreases cell proliferation
  • Reduces ALDH+ cell population size
  • Reduces sphere-formation in colorectal cancer (CRC) cells