Learn More
Medchemexpress LLC Cyclosomatostatin | 84211-54-1 | 99.8% | 779.97 | 1 MG
SDP
Catalog No. 5000424685
Encompass_Preferred
Cyclosomatostatin is a potent somatostatin (SST) receptor antagonist. It can inhibit somatostatin receptor type 1 (SSTR1) signaling and decreases cell proliferation, ALDH+ cell population size, and sphere-formation in colorectal cancer (CRC) cells.
  • Potent somatostatin (SST) receptor antagonist
  • Inhibits somatostatin receptor type 1 (SSTR1) signaling
  • Decreases cell proliferation
  • Decreases ALDH+ cell population size
  • Decreases sphere-formation in colorectal cancer (CRC) cells
  • Target for somatostatin receptor
  • Involved in GPCR/G protein and neuronal signaling pathways
Catalog No. 50-004-24685 Supplier Medchemexpress LLC Supplier No. HYP12011MG
Cyclosomatostatin is a potent somatostatin (SST) receptor antagonist. It can inhibit somatostatin receptor type 1 (SSTR1) signaling and decreases cell proliferation, ALDH+ cell population size, and sphere-formation in colorectal cancer (CRC) cells.
  • Potent somatostatin (SST) receptor antagonist
  • Inhibits somatostatin receptor type 1 (SSTR1) signaling
  • Decreases cell proliferation
  • Decreases ALDH+ cell population size
  • Decreases sphere-formation in colorectal cancer (CRC) cells
  • Target for somatostatin receptor
  • Involved in GPCR/G protein and neuronal signaling pathways