Learn More
Medchemexpress LLC Cyclosomatostatin | 84211-54-1 | 99.8% | 779.97 | 10 MG
SDP
Catalog No. 5000424579
Encompass_Preferred
Cyclosomatostatin is a potent somatostatin (SST) receptor antagonist. It can inhibit somatostatin receptor type 1 (SSTR1) signaling and decreases cell proliferation, ALDH+ cell population size, and sphere-formation in colorectal cancer (CRC) cells.
  • Potent somatostatin (SST) receptor antagonist
  • Inhibits somatostatin receptor type 1 (SSTR1) signaling
  • Decreases cell proliferation in colorectal cancer (CRC) cells
  • Decreases ALDH+ cell population size in colorectal cancer (CRC) cells
  • Decreases sphere-formation in colorectal cancer (CRC) cells
  • White to off-white solid appearance
  • Soluble in H2O (5 mg/mL; needs ultrasonic)
Catalog No. 50-004-24579 Supplier Medchemexpress LLC Supplier No. HYP120110MG
Add to Cart
Add to Cart
Cyclosomatostatin is a potent somatostatin (SST) receptor antagonist. It can inhibit somatostatin receptor type 1 (SSTR1) signaling and decreases cell proliferation, ALDH+ cell population size, and sphere-formation in colorectal cancer (CRC) cells.
  • Potent somatostatin (SST) receptor antagonist
  • Inhibits somatostatin receptor type 1 (SSTR1) signaling
  • Decreases cell proliferation in colorectal cancer (CRC) cells
  • Decreases ALDH+ cell population size in colorectal cancer (CRC) cells
  • Decreases sphere-formation in colorectal cancer (CRC) cells
  • White to off-white solid appearance
  • Soluble in H2O (5 mg/mL; needs ultrasonic)