Learn More
Cayman Chemical Cyclosporin A, 59865-13-3, 100 mg
SDP
Supplier:  Cayman Chemical 12088100
Encompass_Preferred
Molecular Formula C62H111N11O12, Purity ≥99%, Formula Weight 1202.6, Synonyms: NSC 290193; An immunosuppressant drug that binds cyclophilin D, inhibiting the phosphatase activity of calcineurin in T cells (IC50 = 5 nM), which interferes with the nuclear import of NFAT, a transcription factor involved in T cell activation; used for the prevention of transplant rejection.
Catalog No. NC1155210