Learn More
Enzo Life Sciences Cyclosporin A, (100mg), CAS Number: 59865-13-3
SDP
Catalog No. NC9150894
Encompass_Preferred
Immunosuppressant. Alternative Name: CsA. Formula: C62H111N11O12. MW: 1202.6. CAS Number: 59865-13-3. Purity: ≥98% (Assay). Appearance: White to off-white powder. Solubility: Soluble in  DMSO (25mg/ml), methanol, 100% ethanol (25mg/ml), acetone or petroleum ether; insoluble in water. Use/Stability: Store, as supplied, at -20°C for up to 1 year. Store solutions at -20°C for up to 3 months. Long Term Storage: -20°C
Catalog No. NC9150894 Supplier Enzo Life Sciences Supplier No. BMLA1950100
Immunosuppressant. Alternative Name: CsA. Formula: C62H111N11O12. MW: 1202.6. CAS Number: 59865-13-3. Purity: ≥98% (Assay). Appearance: White to off-white powder. Solubility: Soluble in  DMSO (25mg/ml), methanol, 100% ethanol (25mg/ml), acetone or petroleum ether; insoluble in water. Use/Stability: Store, as supplied, at -20°C for up to 1 year. Store solutions at -20°C for up to 3 months. Long Term Storage: -20°C