Learn More
Medchemexpress LLC Cyclosporin A-13C2,d4 | 59865-13-3 | 95.2% | 500 UG
SDP
Catalog No. 5000409030
Encompass_Preferred
Cyclosporin A-13C2,d4 is a 13C labeled Cyclosporin A. This immunosuppressant binds to cyclophilin and inhibits the phosphatase activity of protein phosphatase 2B (PP2B/calcineurin) with an IC50 of 5 nM. It also inhibits CD11a/CD18 adhesion. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during drug development.
  • Functions as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-09030 Supplier Medchemexpress LLC Supplier No. HYB0579S3500UG
Cyclosporin A-13C2,d4 is a 13C labeled Cyclosporin A. This immunosuppressant binds to cyclophilin and inhibits the phosphatase activity of protein phosphatase 2B (PP2B/calcineurin) with an IC50 of 5 nM. It also inhibits CD11a/CD18 adhesion. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during drug development.
  • Functions as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs