Learn More
Medchemexpress LLC HY-B0579 200mg , Cyclosporin A CAS:59865-13-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0579200MG
Encompass_Preferred
HY-B0579 200mg Cyclosporin A CAS: 59865-13-3 Cyclosporin A (Cyclosporine A) is an immunosuppressant which binds to the cyclophilin and inhibits phosphatase activity of calcineurin with an IC50 of 5 nM. Cyclosporin A also inhibits CD11a/CD18 adhesion[8]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9815