Learn More
Medchemexpress LLC HY-B0579 500mg , Cyclosporin A Cyclosporine;Ciclosporin CAS:59865-13-3 Purity:98%
SDP
Catalog No. 501492366
Encompass_Preferred
Medchemexpress, HY-B0579 500mg Cyclosporin A Cyclosporine;Ciclosporin CAS:59865-13-3 Formula:C62H111N11O12 IC50: 7 nM (calcineurin) Purity:98% Cyclosporin A is an immunosuppressive agent binding to the cyclophilin and inhibits calcineurin with an IC 50 of 7 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2366 Supplier Medchemexpress LLC Supplier No. HYB0579500MG
Medchemexpress, HY-B0579 500mg Cyclosporin A Cyclosporine;Ciclosporin CAS:59865-13-3 Formula:C62H111N11O12 IC50: 7 nM (calcineurin) Purity:98% Cyclosporin A is an immunosuppressive agent binding to the cyclophilin and inhibits calcineurin with an IC 50 of 7 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.