Learn More
Selleck Chemical LLC Cyclosporin A S2286-10mM/1mL
SDP
Catalog No. 503112807
Encompass_Preferred
Cyclosporin A a non-polar cyclic oligopeptide is an immunosuppressive agent that binds to cyclophilin and then inhibits calcineurin with IC50 of 7 nM in a cell-free assay Cyclosporin A is generally given following transplant surgery to prevent rejection and has been used to test its toxic effect on a perfused 3D proximal tubule model Cyclosporin A (NSC 290193) can be used to induce animal models of Chronic Rejection of Liver Transplantation
Catalog No. 50-311-2807 Supplier Selleck Chemical LLC Supplier No. S228610MM/1ML
Cyclosporin A a non-polar cyclic oligopeptide is an immunosuppressive agent that binds to cyclophilin and then inhibits calcineurin with IC50 of 7 nM in a cell-free assay Cyclosporin A is generally given following transplant surgery to prevent rejection and has been used to test its toxic effect on a perfused 3D proximal tubule model Cyclosporin A (NSC 290193) can be used to induce animal models of Chronic Rejection of Liver Transplantation