Learn More
Medchemexpress LLC Cyclosporin A-d4 | 59865-13-3 | 99.6% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0579S1MG
Encompass_Preferred
Cyclosporin A-d4 is the deuterium labeled Cyclosporin A. Cyclosporin A (Cyclosporine A) is an immunosuppressant that binds to cyclophilin and inhibits the phosphatase activity of calcineurin with an IC50 of 5 nM. It also inhibits CD11a/CD18 ad.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules, primarily as tracers for quantitation during the drug development process.
  • Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-08381