Learn More
Medchemexpress LLC HY-P1355A 1mg Medchemexpress, Cyclosporin A-Derivative 1 (Free base) CAS:286852-20-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1355A1MG
Encompass_Preferred
Medchemexpress, HY-P1355A 1mg Cyclosporin A-Derivative 1 (Free base) CAS:286852-20-8 Cyclosporin A-Derivative 1 (Free base) is a crystalline intermediate derived from the opening of cyclosporin A extracted from patent WO 2013167703 A1. Cyclosporin A is an immunosuppressive agent which can bind to the cyclophilin and inhibit calcineurin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4189