Learn More
Cayman Chemical Cyclosporin B, Purity: ≥98%, CAS: 63775-95-1, 5 mg
SDP
Catalog No. 502983542
Encompass_Preferred
A fungal metabolite with diverse biological activities; inhibits the growth of T lymphoma cells in a concentration-dependent manner; inhibits P-gp activity in vitro (IC50 = 4.7 μM)
Catalog No. 50-298-3542 Supplier Cayman Chemical Supplier No. 195255 MG
A fungal metabolite with diverse biological activities; inhibits the growth of T lymphoma cells in a concentration-dependent manner; inhibits P-gp activity in vitro (IC50 = 4.7 μM)