Learn More
Cayman Chemical CyclosporIn C 10mg
SDP
Catalog No. 502939678
Encompass_Preferred
A fungal metabolite with diverse biological activities; active against isolates of B. cinerea, A. niger, and Alternaria, Mucor, and Penicillium species (MICs = 0.1-5 μg/ml); inhibits vaccinia virus replication in infected BSC40 cells by 98.82% at 15 μg/ml; inhibits lymphocyte proliferation induced by the mitogens ConA, PHA, and PWM, as well as proliferation induced by alloantigen in mixed lymphocyte at 100 ng/ml; inhibits the local GVH reaction in mice receiving splenocyte grafts at 100 ng/ml
Catalog No. 50-293-9678 Supplier Cayman Chemical Supplier No. 2915210 MG
A fungal metabolite with diverse biological activities; active against isolates of B. cinerea, A. niger, and Alternaria, Mucor, and Penicillium species (MICs = 0.1-5 μg/ml); inhibits vaccinia virus replication in infected BSC40 cells by 98.82% at 15 μg/ml; inhibits lymphocyte proliferation induced by the mitogens ConA, PHA, and PWM, as well as proliferation induced by alloantigen in mixed lymphocyte at 100 ng/ml; inhibits the local GVH reaction in mice receiving splenocyte grafts at 100 ng/ml