Learn More
Enzo Life Sciences Cyclosporin C (5mg). CAS: 59787-61-0
SDP
Supplier:  Enzo Life Sciences ALX380282M005
Encompass_Preferred
Potent immunosuppressor. Alternative name: Thr2-cyclosporin A, 7-L-Threonine-cyclosporin A. Purity: ≥95% (HPLC). Appearance: Colorless to off-white crystalline solid. Solubility: Soluble in 100% ethanol, acetone or ethyl acetate; insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-9012