Learn More
Enzo Life Sciences Cyclosporin D (5mg). CAS: 63775-96-2
SDP
Supplier:  Enzo Life Sciences ALX380284M005
Encompass_Preferred
Weak immunosuppressor. Potent inhibitor of tumor promoting phorbol ester TPA/PMA (BML-PE160) in vivo. Potent inhibitor of Ca2+/calmodulin dependent EF-2 phosphorylation in vitro. Alternative name: Val2-cyclosporin, 7-L-valine-Cyclosporin A. Purity: ≥95% (HPLC). Solubility: Soluble in 100% ethanol, acetone or ethyl acetate; insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-9014