Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclosporin D | 63775-96-2 | 1216.64 | C63H113N11O12 | 5 MG
SDP

Catalog No. 5000301462
Encompass_Preferred

Cyclosporin D is a cyclic peptide metabolite of cyclosporin A supplied as an analytical standard for research and quantitative assays. It is provided as a powder (available in 1 mg, 5 mg, and 10 mg sizes), has molecular formula C63H113N11O12 and molecular weight 1216.64 Da, and requires sealed, low-temperature storage to maintain stability.

  • Cyclic peptide metabolite used as an analytical standard.
  • Molecular formula C63H113N11O12; molecular weight 1216.64 Da.
  • Available in small vial pack sizes for analytical use.
  • Powder form with sealed, moisture-protected storage recommended.
  • Storage stability: powder -80°C (2 years) and -20°C (1 year); in solvent -80°C (6 months) and -20°C (1 month).
  • Intended for research and quantitative assay applications.

Catalog No. 50-003-01462 Supplier Medchemexpress LLC Supplier No. HYW0197215MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclosporin D is a cyclic peptide metabolite of cyclosporin A supplied as an analytical standard for research and quantitative assays. It is provided as a powder (available in 1 mg, 5 mg, and 10 mg sizes), has molecular formula C63H113N11O12 and molecular weight 1216.64 Da, and requires sealed, low-temperature storage to maintain stability.

  • Cyclic peptide metabolite used as an analytical standard.
  • Molecular formula C63H113N11O12; molecular weight 1216.64 Da.
  • Available in small vial pack sizes for analytical use.
  • Powder form with sealed, moisture-protected storage recommended.
  • Storage stability: powder -80°C (2 years) and -20°C (1 year); in solvent -80°C (6 months) and -20°C (1 month).
  • Intended for research and quantitative assay applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.