Learn More
Medchemexpress LLC Cyclosporin D | 63775-96-2 | MFCD11111526 | 99.9% | 1,216.64 g/mol | C63H113N11O12 | 10 MG
SDP
Catalog No. 5000300867
Encompass_Preferred
Cyclosporin D is a cyclic undecapeptide and a metabolite/analogue of cyclosporin A supplied as a research standard for analytical, biochemical, and pharmacological studies. It is commonly used as an internal standard and reference material for assay development and quantification.
  • Cyclic undecapeptide structure suitable for biochemical studies.
  • High purity for analytical applications.
  • Available in small vial sizes for research use.
  • Useful as an internal standard for quantification of cyclosporin A.
  • Stable when stored according to supplier recommendations.
Catalog No. 50-003-00867 Supplier Medchemexpress LLC Supplier No. HYW01972110MG
Cyclosporin D is a cyclic undecapeptide and a metabolite/analogue of cyclosporin A supplied as a research standard for analytical, biochemical, and pharmacological studies. It is commonly used as an internal standard and reference material for assay development and quantification.
  • Cyclic undecapeptide structure suitable for biochemical studies.
  • High purity for analytical applications.
  • Available in small vial sizes for research use.
  • Useful as an internal standard for quantification of cyclosporin A.
  • Stable when stored according to supplier recommendations.