Learn More
AdipoGen Cyclosporin D
SDP
Catalog No. 502562524
Encompass_Preferred
Chemical. CAS 63775-96-2. Formula C63H113N11O12. MW 1216.7. Isolated from Fusarium solani. Potent inhibitor of tumor promoting phorbol ester TPA/PMA AG-CN2-0010 effects in vivo. Antifungal. Moderate immunosuppressant. Calcineurin inhibitor with lower activity than Cyclosporin A AG-CN2-0079. Moderate inhibitor of formyl peptide-induced superoxide formation.
Catalog No. 50-256-2524 Supplier AdipoGen Supplier No. AGCN20445M005
Chemical. CAS 63775-96-2. Formula C63H113N11O12. MW 1216.7. Isolated from Fusarium solani. Potent inhibitor of tumor promoting phorbol ester TPA/PMA AG-CN2-0010 effects in vivo. Antifungal. Moderate immunosuppressant. Calcineurin inhibitor with lower activity than Cyclosporin A AG-CN2-0079. Moderate inhibitor of formyl peptide-induced superoxide formation.