Learn More
Enzo Life Sciences Cyclosporin H, (1mg), CAS Number: 83602-39-5
SDP
Supplier:  Enzo Life Sciences ALX380286M001
Encompass_Preferred
TPA/PMA inhibitor. Immunologically inactive. Does not bind to immunophilin. Potent inhibitor of tumor promoting phorbol ester TPA/PMA (Prod. No. BML-PE160) in mouse skin in vivo. Potent inhibitor of Ca2+/calmodulin dependent EF-2 phosphorylation in vitro. Potent and selective antagonist of formyl peptide receptor. Inhibits formyl peptide-induced superoxide formation. . Alternative Name: Csh-cyclosporin, 5-(N-Methyl-D-valine)-cyclosporin A. Formula: C62H111N11O12. MW: 1202.6. CAS Number: 83602-39-5. Purity: ≥95% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in 100% ethanol, acetone or ethyl acetate; insoluble in water. Long Term Storage: +4°C
Catalog No. NC9767496