Learn More
AdipoGen Cyclosporin H (5 mg)
SDP
Catalog No. 502053614
Encompass_Preferred
Chemical. CAS: 83602-39-5. Formula: C62H111N11O12. MW: 1202.6. Isolated from Tolypocladium inflatum. Potent inhibitor of tumor promoting phorbol ester TPA/PMA (AG-CN2-0010) effects in vivo. Antifungal. Potent and effective inhibitor of formyl peptide receptor (FPR) and formyl peptide-induced superoxide formation. Shows no immunosuppresive activity like other cyclosporins.
Catalog No. 50-205-3614 Supplier AdipoGen Supplier No. AGCN20447M005
Chemical. CAS: 83602-39-5. Formula: C62H111N11O12. MW: 1202.6. Isolated from Tolypocladium inflatum. Potent inhibitor of tumor promoting phorbol ester TPA/PMA (AG-CN2-0010) effects in vivo. Antifungal. Potent and effective inhibitor of formyl peptide receptor (FPR) and formyl peptide-induced superoxide formation. Shows no immunosuppresive activity like other cyclosporins.