Learn More
Medchemexpress LLC HY-P1122 5mg Medchemexpress, Cyclosporin H CAS:83602-39-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP11225MG
Encompass_Preferred
Medchemexpress, HY-P1122 5mg Cyclosporin H CAS:83602-39-5 Cyclosporin H, a viral transduction enhancer, increases lentiviral transduction up to 10-fold in human cord blood-derived hematopoietic stem and progenitor cells (HSPCs). Cyclosporin H displays an additive effect when combined with Rapamycin (HY-10219) or Prostaglandin E2 (HY-101952). Cyclosporin H lacks immunosuppressant activity of Cyclosporin A. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3039