Learn More
Abcam Cyclosporin H, TPA/PMA inhibitor, 1MG
Catalog No. 502706400
Encompass_Preferred
MW 1216.6 Da, Purity >96%. TPA/PMA inhibitor. Selective inhibitor of formyl peptide receptors (Ki = 0.1 μM). Inhibits Ca²+/calmodulin dependent EF-2 phosphorylation. Inhibits FMLP-induced superoxide anion formation (IC₅₀ = 40 nM). Multiple biological effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6400 Supplier Abcam Supplier No. AB1419231MG
MW 1216.6 Da, Purity >96%. TPA/PMA inhibitor. Selective inhibitor of formyl peptide receptors (Ki = 0.1 μM). Inhibits Ca²+/calmodulin dependent EF-2 phosphorylation. Inhibits FMLP-induced superoxide anion formation (IC₅₀ = 40 nM). Multiple biological effects.
The product is subject to the following: Abcam Restricted Use Statement