Learn More
TOKU-E Cyclosporin A ReadyMade™ Solution | CAS 59865-13-3 | 5 mg/ML in DMSO | sterile-filtered | 10 x 1 ML
Catalog No. NC2068741
Encompass_Preferred
Cyclosporin A ReadyMade™ Solution | CAS 59865-13-3 | 5 mg/ML in DMSO | sterile-filtered | 10 x 1 ML
Catalog No. NC2068741 Supplier TOKU-E Supplier No. C28510X1ML
Cyclosporin A ReadyMade™ Solution | CAS 59865-13-3 | 5 mg/ML in DMSO | sterile-filtered | 10 x 1 ML