Learn More
Selleck Chemical LLC Cyclosporine 50mg 79217-60-0
SDP
Supplier:  Selleck Chemical LLC S151450MG
Encompass_Preferred
Cyclosporine is a calcineurin phosphatase pathway inhibitor, used as an immunosuppressant drug to prevent rejection in organ transplantation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-734-0