Learn More
Foss™ Cyclotec™ Sample Mill Replacement Seals
Catalog No. TC10001990
Change view
Click to view available options
For Use With (Equipment):
For sample bottle
Catalog No. For Use With (Equipment)
TC1000-1990 For sample bottle
1 options
Catalog No. TC1000-1990 Supplier Foss™ Supplier No. 10001990
For Cyclotec 1093 and Cyclotec 193, Cannot be used with the Cyclotec 293.
Specifications
Type Seal
For Use With (Equipment) For sample bottle