Learn More
Cayman Chemical Cyclothiazide, Purity: ≥95%, CAS: 2259-96-3, 50 mg
SDP
Catalog No. 502970542
Encompass_Preferred
A benzothiadiazide that acts as a potentiator of AMPA receptors, positively modulating its response to glutamic acid (EC50 = 3.8 µM); also a negative modulator of GABAA receptors, reversibly inhibiting both evoked and spontaneous inhibitory postsynaptic currents (IC50 = 58 µM)
Catalog No. 50-297-0542 Supplier Cayman Chemical Supplier No. 1633550 MG
A benzothiadiazide that acts as a potentiator of AMPA receptors, positively modulating its response to glutamic acid (EC50 = 3.8 µM); also a negative modulator of GABAA receptors, reversibly inhibiting both evoked and spontaneous inhibitory postsynaptic currents (IC50 = 58 µM)