Learn More
Abcam Cyclothiazide, AMPA receptor desensitisation inhibitor, 10MG
Catalog No. 502707895
Encompass_Preferred
MW 389.9 Da, Purity >98%. Positive allosteric modulator of AMPA receptors. Produces a fast inhibition of AMPA receptor desensitization and a much slower potentiation of the AMPA current. Also available in Kit: Ionotropic agonists (ab120323).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7895 Supplier Abcam Supplier No. AB12006110MG
MW 389.9 Da, Purity >98%. Positive allosteric modulator of AMPA receptors. Produces a fast inhibition of AMPA receptor desensitization and a much slower potentiation of the AMPA current. Also available in Kit: Ionotropic agonists (ab120323).
The product is subject to the following: Abcam Restricted Use Statement