Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclotridecanone, Tech., Thermo Scientific™

Catalog No. AC171020025
Change view
Click to view available options
Quantity:
2.5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC171020025 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC171020025 Supplier Thermo Scientific Chemicals Supplier No. 171020025

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 832-10-0
Molecular Formula C13H24O
Molecular Weight (g/mol) 196.334
InChI Key VHUGWUBIUBBUAF-UHFFFAOYSA-N
Synonym cyclotridecanon, cyclotridecan-1-one, cyclotridecane-1-one, acmc-20albq
PubChem CID 79114
IUPAC Name cyclotridecanone
SMILES C1CCCCCCC(=O)CCCCC1

Specifications

Linear Formula C13H24(=O)
Quantity 2.5 g
Beilstein 07, II, 49
Formula Weight 196.33
Grade Technical
Chemical Name or Material Cyclotridecanone, tech.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.