Learn More
AdipoGen Cyclotrisiloxanol pentamethyl
SDP
Catalog No. 502561343
Encompass_Preferred
Chemical. CAS 106916-50-1. Formula C5H16O4Si3. MW 224.43. Organosilicon compounds are organometallic compounds containing carbon-silicon bonds. Organosilicon compounds are widely encountered in commercial products. Most common are antifoamers, caulks sealant, adhesives, and coatings made from silicones. Other important uses include agricultural and plant control adjuvants commonly used in conjunction with herbicides and fungicides. Several organosilicon compounds have been investigated as pharmaceuticals.
Catalog No. 50-256-1343 Supplier AdipoGen Supplier No. CDXC0753G001
Chemical. CAS 106916-50-1. Formula C5H16O4Si3. MW 224.43. Organosilicon compounds are organometallic compounds containing carbon-silicon bonds. Organosilicon compounds are widely encountered in commercial products. Most common are antifoamers, caulks sealant, adhesives, and coatings made from silicones. Other important uses include agricultural and plant control adjuvants commonly used in conjunction with herbicides and fungicides. Several organosilicon compounds have been investigated as pharmaceuticals.