Learn More
Medchemexpress LLC HY-N0107 5mg Medchemexpress, Cyclovirobuxine D CAS:860-79-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN01075MG
Encompass_Preferred
Medchemexpress, HY-N0107 5mg Cyclovirobuxine D CAS:860-79-7 Cyclovirobuxine D (CVB-D) is the main active component of the traditional Chinese medicine Buxus microphylla. Cyclovirobuxine D induces autophagy and attenuates the phosphorylation of Akt and mTOR. Cyclovirobuxine D inhibits cell proliferation of gastric cancer cells through suppression of cell cycle progression and inducement of mitochondria-mediated apoptosis. Cyclovirobuxine D is beneficial for heart failure induced by myocardial infarction[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0223