Learn More
Medchemexpress LLC HY-111653 10mg Medchemexpress, CycLuc1 CAS:1247879-16-8 Purity:>98%
SDP
Catalog No. 501968937
Encompass_Preferred
Medchemexpress, HY-111653 10mg CycLuc1 CAS:1247879-16-8 CycLuc1 is a brain penetrant luciferase substrate. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8937 Supplier Medchemexpress LLC Supplier No. HY11165310MG
Medchemexpress, HY-111653 10mg CycLuc1 CAS:1247879-16-8 CycLuc1 is a brain penetrant luciferase substrate. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.