Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 54205MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: 7H8.2C12. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b kappa. Positive Control: K-562, HL-60, Jurkat, NIH3T3 or PC-3 cells. Liver or Cardiac muscle. Cellular Staining: Cytoplasmic. Buffer: 1mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Synthetic peptides corresponding to amino acid 1-80, 81-104, 66-104 of pigeon cytochrome c. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 15kDa.Chromosome: 7p15.3. Gene Symbol: CYCS. Gene ID: 54205. Ensemble: ENSG00000172115. Uniprot: NP_061820.
Catalog No. 50-188-5056