Learn More
FUJIFILM Biosciences Cyenopyrafen Standard
Catalog No. 5000450339
Encompass_Preferred
Cyenopyrafen Standard is a high-purity analytical reference material 98 0% developed for pesticide residue analysis This compound is ideal for use in HPLC and quantitative NMR qNMR methods due to its high assay values and stability under proper storage conditions 2 10 C protected from light in inert atmosphere
Catalog No. 50-004-50339 Supplier FUJIFILM Biosciences Supplier No. 03022681
Cyenopyrafen Standard is a high-purity analytical reference material 98 0% developed for pesticide residue analysis This compound is ideal for use in HPLC and quantitative NMR qNMR methods due to its high assay values and stability under proper storage conditions 2 10 C protected from light in inert atmosphere