Learn More
Aviva Systems Biology Corp CYFIP1 Antibody - C-terminal region (ARP64537_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64537_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYFIP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human CYFIP1. Alternate names: FLJ45151, P140SRA-1, SHYC, SRA1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-291