Learn More
ABclonal Technology CYFIP1 Rabbit mAb
SDP
Catalog No. 5000008002
Encompass_Preferred
This gene encodes a protein that regulates cytoskeletal dynamics and protein translation The encoded protein is a component of the WAVE regulatory complex (WRC) which promotes actin polymerization This protein also interacts with the synaptic functional regulator FMR1 protein and translation initiation factor 4E to inhibit protein translation A large chromosomal deletion including this gene is associated with increased risk of schizophrenia and epilepsy in human patients Reduced expression of this gene has been observed in various human cancers and the encoded protein may inhibit tumor invasion
Catalog No. 50-000-08002 Supplier ABclonal Technology Supplier No. A2092420UL
This gene encodes a protein that regulates cytoskeletal dynamics and protein translation The encoded protein is a component of the WAVE regulatory complex (WRC) which promotes actin polymerization This protein also interacts with the synaptic functional regulator FMR1 protein and translation initiation factor 4E to inhibit protein translation A large chromosomal deletion including this gene is associated with increased risk of schizophrenia and epilepsy in human patients Reduced expression of this gene has been observed in various human cancers and the encoded protein may inhibit tumor invasion