Learn More
FUJIFILM Biosciences Cyflufenamid Standard
Catalog No. 5000450335
Encompass_Preferred
Cyflufenamid Standard is a highly pure analytical reference material for pesticide residue analysis in environmental and agricultural samples It ensures accurate quantification using GC or qNMR methods
Catalog No. 50-004-50335 Supplier FUJIFILM Biosciences Supplier No. 03619231
Cyflufenamid Standard is a highly pure analytical reference material for pesticide residue analysis in environmental and agricultural samples It ensures accurate quantification using GC or qNMR methods